Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Erlose, MP Biomedicals™

Catalog No. ICN19866883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19866883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19866883 Supplier MP Biomedicals, Inc Supplier No. 0219866883
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13101-54-7
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00190706
InChI Key FVVCFHXLWDDRHG-KKNDGLDKSA-N
Synonym erlose, glucosylsucrose, sucrosylglucose, beta-d-fructofuranosyl alpha-d-glucopyranosyl-1->4-alpha-d-glucopyranoside, alpha-maltosyl beta-fructofuranoside, alpha-d-glc-1->4-alpha-d-glc-1->2-beta-d-fru, a-d-glucopyranoside, b-d-fructofuranosyl o-a-d-glucopyranosyl-1, alpha-d-glucopyranoside, beta-d-fructofuranosyl o-alpha-d-glucopyranosyl-1-4, alpha-d-glucopyranosyl-1,2,3,4-alpha-d-glucopyranosyl-1,2-beta-d-fructofuranoside, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-2s,3s,4s,5r-3,4-dihydroxy-2,5-bis hydroxymethyl oxolan-2-yl oxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol
PubChem CID 193471
ChEBI CHEBI:55512
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3(C(C(C(O3)CO)O)O)CO)CO)O)O)O)O

Specifications

Color White
Quantity 250 mg
Formula Weight 504.4
Physical Form Crystalline Powder
Chemical Name or Material Erlose
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.