Learn More
Selleck Chemical LLC Erlotinib 200mg 183321-74-6 CP358774, NSC 718781
SDP
Supplier:  Selleck Chemical LLC S7786200MG
Encompass_Preferred
Erlotinib (CP358774, NSC 718781) is an EGFR inhibitor with IC50 of 2 nM, >1000-fold more sensitive for EGFR than human c-Src or v-Abl. Erlotinib induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1234640