Learn More
Medchemexpress LLC HY-50896 500mg , Erlotinib NSC 718781;OSI-744;R1415 CAS:183321-74-6 Purity:98%
SDP
Catalog No. 501492338
Encompass_Preferred
Medchemexpress, HY-50896 500mg Erlotinib NSC 718781;OSI-744;R1415 CAS:183321-74-6 Formula:C22H23N3O4 IC50: 2 nM (EGFR) Purity:98% Erlotinib inhibits purified EGFR kinase with an IC 50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2338 Supplier Medchemexpress LLC Supplier No. HY50896500MG
Medchemexpress, HY-50896 500mg Erlotinib NSC 718781;OSI-744;R1415 CAS:183321-74-6 Formula:C22H23N3O4 IC50: 2 nM (EGFR) Purity:98% Erlotinib inhibits purified EGFR kinase with an IC 50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.