Learn More
Medchemexpress LLC Erlotinib-d6 hydrochloride | 1189953-78-3 | 98.1% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12008S1MG
Encompass_Preferred
Erlotinib-d6 hydrochloride is a deuterium-labeled form of erlotinib hydrochloride used as an internal standard and analytical reagent for studies targeting the epidermal growth factor receptor (EGFR). Supplied as the hydrochloride salt to aid solubility, it is intended for research use only.
  • Deuterium-labeled internal standard for quantitative analysis
  • High purity suitable for LC-MS and analytical assays
  • Molecular formula C22H18D6ClN3O4, molecular weight 435.93
  • Available in 1 MG packs for small-scale experiments
  • Intended for research use only; not for human consumption or clinical use
Catalog No. 50-002-32006