Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib-d6 hydrochloride | 1189953-78-3 | 98.1% | 10MG
SDP

Catalog No. 5000233491
Encompass_Preferred

Erlotinib-d6 hydrochloride | 1189953-78-3 | 98.1% | 10MG

Catalog No. 50-002-33491 Supplier Medchemexpress LLC Supplier No. HY12008S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erlotinib-d6 hydrochloride | 1189953-78-3 | 98.1% | 10MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.