Learn More
Medchemexpress LLC Erlotinib-d6 hydrochloride | 1189953-78-3 | 98.1% | 5 MG
SDP
Catalog No. 5000233406
Encompass_Preferred
Deuterated erlotinib hydrochloride (d6) is a deuterium-labeled analytical standard used as an internal standard for LC-MS quantification and pharmacokinetic studies. Supplied as a powder, it provides defined isotopic labeling for accurate tracer and quantitation workflows.
  • Deuterated (d6) analogue for internal standard use.
  • High isotopic labeling for reliable quantitation.
  • Powder form, suitable for solution preparation.
  • Soluble in water ≥ 0.1 mg/mL (0.23 mM) under in vitro conditions.
  • Powder stable at -20°C for up to 3 years; in solvent stability varies.
Catalog No. 50-002-33406 Supplier Medchemexpress LLC Supplier No. HY12008S5MG
Deuterated erlotinib hydrochloride (d6) is a deuterium-labeled analytical standard used as an internal standard for LC-MS quantification and pharmacokinetic studies. Supplied as a powder, it provides defined isotopic labeling for accurate tracer and quantitation workflows.
  • Deuterated (d6) analogue for internal standard use.
  • High isotopic labeling for reliable quantitation.
  • Powder form, suitable for solution preparation.
  • Soluble in water ≥ 0.1 mg/mL (0.23 mM) under in vitro conditions.
  • Powder stable at -20°C for up to 3 years; in solvent stability varies.