Learn More
Apexbio Technology LLC Erlotinib Hydrochloride 183319-69-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062674
Encompass_Preferred
Erlotinib hydrochloride (CAS 183319-69-9) is a small molecule inhibitor targeting the epidermal growth factor receptor (EGFR/HER-1) specifically blocking its tyrosine kinase activity By competitively inhibiting EGFR phosphorylation (IC50 2 nM) Erlotinib hydrochloride disrupts downstream signaling pathways involved in tumor cell proliferation apoptosis resistance migration and angiogenesis In vitro studies indicate this inhibitor induces G0/G1 cell cycle arrest and promotes apoptosis in cancer cell lines such as pancreatic (Bxpc-3 PANC-1) and hepatocellular carcinoma cells Erlotinib hydrochloride is widely studied in oncology research particularly in non-small cell lung cancer (NSCLC) and pancreatic cancer models
Catalog No. 50-000-62674 Supplier Apexbio Technology LLC Supplier No. A82345.1
Erlotinib hydrochloride (CAS 183319-69-9) is a small molecule inhibitor targeting the epidermal growth factor receptor (EGFR/HER-1) specifically blocking its tyrosine kinase activity By competitively inhibiting EGFR phosphorylation (IC50 2 nM) Erlotinib hydrochloride disrupts downstream signaling pathways involved in tumor cell proliferation apoptosis resistance migration and angiogenesis In vitro studies indicate this inhibitor induces G0/G1 cell cycle arrest and promotes apoptosis in cancer cell lines such as pancreatic (Bxpc-3 PANC-1) and hepatocellular carcinoma cells Erlotinib hydrochloride is widely studied in oncology research particularly in non-small cell lung cancer (NSCLC) and pancreatic cancer models