Learn More
Selleck Chemical LLC Erlotinib HCl (OSI-744) 100mg 183319-69-9 CP358774, NSC 718781
SDP
Supplier:  Selleck Chemical LLC S1023100MG
Encompass_Preferred
Erlotinib HCl (OSI-744, CP358774, NSC 718781) is an EGFR inhibitor with IC50 of 2 nM in cell-free assays, >1000-fold more sensitive for EGFR than human c-Src or v-Abl. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0514936