Learn More
Selleck Chemical LLC Erlotinib HCl (OSI-744) 500mg 183319-69-9 CP358774, NSC 718781
SDP
Catalog No. 506203
Encompass_Preferred
Erlotinib HCl (OSI-744, CP358774, NSC 718781) is an EGFR inhibitor with IC50 of 2 nM in cell-free assays, >1000-fold more sensitive for EGFR than human c-Src or v-Abl. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-620-3 Supplier Selleck Chemical LLC Supplier No. S1023500MG
Erlotinib HCl (OSI-744, CP358774, NSC 718781) is an EGFR inhibitor with IC50 of 2 nM in cell-free assays, >1000-fold more sensitive for EGFR than human c-Src or v-Abl. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.