Learn More
Medchemexpress LLC HY-12008 500mg , Erlotinib (Hydrochloride) CP-358774;OSI-774;NSC 718781 CAS:183319-69-9 Purity:98%
SDP
Catalog No. 501492461
Encompass_Preferred
Medchemexpress, HY-12008 500mg Erlotinib (Hydrochloride) CP-358774;OSI-774;NSC 718781 CAS:183319-69-9 Formula:C22H24ClN3O4 IC50: 2 nM (EGFR) Purity:98% Erlotinib hydrochloride inhibits purified EGFR kinase with an IC 50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2461 Supplier Medchemexpress LLC Supplier No. HY12008500MG
Medchemexpress, HY-12008 500mg Erlotinib (Hydrochloride) CP-358774;OSI-774;NSC 718781 CAS:183319-69-9 Formula:C22H24ClN3O4 IC50: 2 nM (EGFR) Purity:98% Erlotinib hydrochloride inhibits purified EGFR kinase with an IC 50 of 2 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.