Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib hydrochloride | 183319-69-9 | >99.0% | 10 G
SDP

Catalog No. 5000234109
Encompass_Preferred

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, a potent epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used as a research reagent. It inhibits purified EGFR kinase (IC50 ≈ 2 nM) and shows anti-tumor activity in preclinical studies. For research use only.

  • Inhibits EGFR kinase (IC50 ≈ 2 nM).
  • Molecular weight 429.90 g/mol.
  • Chemical formula C22H24ClN3O4.
  • Purity >99% (manufacturer claim).
  • Supplied as an off-white powder; store at -20°C.

Catalog No. 50-002-34109 Supplier Medchemexpress LLC Supplier No. HY1200810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, a potent epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor used as a research reagent. It inhibits purified EGFR kinase (IC50 ≈ 2 nM) and shows anti-tumor activity in preclinical studies. For research use only.

  • Inhibits EGFR kinase (IC50 ≈ 2 nM).
  • Molecular weight 429.90 g/mol.
  • Chemical formula C22H24ClN3O4.
  • Purity >99% (manufacturer claim).
  • Supplied as an off-white powder; store at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.