Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib hydrochloride | 183319-69-9 | 99.7% | 25 MG
SDP

Supplier:  Medchemexpress LLC HY12008R25MG

Encompass_Preferred

Erlotinib hydrochloride analytical standard, the hydrochloride salt of erlotinib, provided as a high-purity solid for use as a reference material in biochemical and analytical assays.

  • Analytical standard for erlotinib hydrochloride.
  • Purity 99.73% (analytical).
  • Molecular weight 429.90 g/mol.
  • Chemical formula C22H24ClN3O4.
  • Appearance: white to off-white solid.
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg; 200 mg and 500 mg available by quote.
  • Store under conditions specified on the certificate of analysis.

Catalog No. 50-002-31860


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.