Learn More
Medchemexpress LLC Erlotinib hydrochloride | 183319-69-9 | 99.7% | 5 MG

Supplier: Medchemexpress LLC HY12008R5MG
Erlotinib hydrochloride is the hydrochloride salt of erlotinib, a quinazoline-derived epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor supplied as an analytical standard for research and analytical applications. It is presented in small pack sizes suitable for laboratory assays.
- Analytical standard for EGFR inhibitor studies.
- CAS number 183319-69-9.
- Purity 99.7% (analytical grade).
- Molecular formula C22H24ClN3O4.
- Molecular weight 429.90 g/mol.
- Available in 5 mg pack size for small-scale laboratory use.
- Suitable as a reference standard for analytical assays.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.