Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib (hydrochloride) | 183319-69-9 | 99.9% | 1 G
SDP

Catalog No. 5000233323
Encompass_Preferred

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase intended for research use. It inhibits purified EGFR kinase with an IC50 of 2 nM and contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (click chemistry).

  • Inhibits EGFR kinase; IC50 = 2 nM.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High purity (99.94%) suitable for analytical and research applications.
  • Molecular formula C22H24ClN3O4, molecular weight 429.90.
  • Solid, melting point 223-225 °C; recommended storage 4 °C (sealed).
  • Typically supplied as a 1 g research pack.

Catalog No. 50-002-33323 Supplier Medchemexpress LLC Supplier No. HY120081G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase intended for research use. It inhibits purified EGFR kinase with an IC50 of 2 nM and contains an alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (click chemistry).

  • Inhibits EGFR kinase; IC50 = 2 nM.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • High purity (99.94%) suitable for analytical and research applications.
  • Molecular formula C22H24ClN3O4, molecular weight 429.90.
  • Solid, melting point 223-225 °C; recommended storage 4 °C (sealed).
  • Typically supplied as a 1 g research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.