Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib hydrochloride | 183319-69-9 | 99.9% | 10 MG
SDP

Catalog No. 5000234077
Encompass_Preferred

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, an EGFR tyrosine kinase inhibitor used in research to study oncology, signal transduction, and inhibitor profiling. Supplied as a high-purity solid suitable for in vitro assays and formulation development.

  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 429.90 g/mol; formula C22H24ClN3O4.
  • Soluble in DMSO (~3.33 mg/mL); in vivo formulation guidance provided by manufacturer.
  • Available in small-mass packages for assay development, including 10 mg size.
  • Acts as an EGFR tyrosine kinase inhibitor (IC50 ≈ 2 nM) for target validation studies.

Catalog No. 50-002-34077 Supplier Medchemexpress LLC Supplier No. HY12008R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erlotinib hydrochloride is the hydrochloride salt of erlotinib, an EGFR tyrosine kinase inhibitor used in research to study oncology, signal transduction, and inhibitor profiling. Supplied as a high-purity solid suitable for in vitro assays and formulation development.

  • High purity suitable for biochemical and cell-based assays.
  • Molecular weight 429.90 g/mol; formula C22H24ClN3O4.
  • Soluble in DMSO (~3.33 mg/mL); in vivo formulation guidance provided by manufacturer.
  • Available in small-mass packages for assay development, including 10 mg size.
  • Acts as an EGFR tyrosine kinase inhibitor (IC50 ≈ 2 nM) for target validation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.