Learn More
Apexbio Technology LLC Erlotinib Hydrochloride, 183319-69-9, MFCD07781272, 1 g
SDP
Supplier:  Apexbio Technology LLC A82341000
Encompass_Preferred
MF: C22H24ClN3O4, MW: 429.91, Solubility: 6.44 mg/mL in DMSO with gentle warming,insoluble in EtOH,insoluble in H2O. Erlotinib hydrochloride is a directly acting inhibitor of epidermal growth factor receptor (EGFR/HER-1) tyrosine kinase with an IC50 of 2 nM.
Catalog No. 50-101-2655