Learn More
Apexbio Technology LLC Erlotinib Hydrochloride, 5g. Cas: 183319-69-9 MFCD: MFCD07781272. Selective EGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A82345000
Encompass_Preferred
Erlotinib hydrochloride (the trade name Tarceva?) is a directly acting inhibitor of epidermal growth factor receptor (EGFR/HER-1) tyrosine kinase with an IC50 of 2 nM.Epidermal growth factor receptor (EGFR) is one member of the ErbB family which includes EGFR (ErbB1), ErbB2, ErbB3 and ErbB4. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5087