Learn More
Abcam Erlotinib hydrochloride, EGFR tyrosine kinase inhibitor, 100MG
Catalog No. 502706399
Encompass_Preferred
MW 429.9 Da, Purity >98%. Potent EGFR tyrosine kinase inhibitor (IC₅₀ = 2 nM). Decreases EGFR autophosphorylation in tumor cells. Potent inhibitor of mutant JAK2-V617F. Weaker inhibitor of ErbB-2 kinase (Ki = 1 μM) and ErbB-4 kinase (Ki = 1.5 μM). Clinically effective agent for non-small cell lung cancer (NSCLC).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6399 Supplier Abcam Supplier No. AB141930100MG
MW 429.9 Da, Purity >98%. Potent EGFR tyrosine kinase inhibitor (IC₅₀ = 2 nM). Decreases EGFR autophosphorylation in tumor cells. Potent inhibitor of mutant JAK2-V617F. Weaker inhibitor of ErbB-2 kinase (Ki = 1 μM) and ErbB-4 kinase (Ki = 1.5 μM). Clinically effective agent for non-small cell lung cancer (NSCLC).
The product is subject to the following: Abcam Restricted Use Statement