Learn More
Thermo Scientific Chemicals Erlotinib hydrochloride
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 215.0°C to 227.0°C |
| Color | Clear |
| Quantity | 1 g |
| Solubility Information | Solubility aq. soln.: slightly soluble. Other solubilities: soluble in dimethylsulfoxide, slightly soluble in ethanol |
| Formula Weight | 429.9 |
| Chemical Name or Material | Erlotinib hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.