Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erlotinib impurity 44 | 183321-85-9 | >98.0% | 1 MG
SDP

Catalog No. 5000233492
Encompass_Preferred

Erlotinib impurity 44 is an impurity-related small molecule used as an analytical reference or synthetic intermediate in medicinal chemistry research. It is supplied for research use and is typically handled and stored under refrigerated, inert conditions.

  • Cas 183321-85-9 for unambiguous identification.
  • Molecular formula C21H20ClN3O3 and molecular weight 397.85 g·mol⁻¹.
  • Purity reported as greater than 98% for analytical and reference use.
  • Stored at 4 °C under nitrogen to preserve stability.
  • Intended for impurity profiling, method development, and synthetic chemistry research.

Catalog No. 50-002-33492 Supplier Medchemexpress LLC Supplier No. HY12008Z1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Erlotinib impurity 44 is an impurity-related small molecule used as an analytical reference or synthetic intermediate in medicinal chemistry research. It is supplied for research use and is typically handled and stored under refrigerated, inert conditions.

  • Cas 183321-85-9 for unambiguous identification.
  • Molecular formula C21H20ClN3O3 and molecular weight 397.85 g·mol⁻¹.
  • Purity reported as greater than 98% for analytical and reference use.
  • Stored at 4 °C under nitrogen to preserve stability.
  • Intended for impurity profiling, method development, and synthetic chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.