Learn More
Cayman Chemical ErlotInIbhydrochlorIde 50mg
SDP
Catalog No. 502913698
Encompass_Preferred
An EGFR inhibitor (Ki = 2.7 nM); selective for EGFR over Src and Abl at 0.1 µM; induces caspase-3 and -7 activity in NCI H358 cells at 1 µM; reduces EGFR autophosphorylation in an HN-5 mouse xenograft model at 25 mg/kg
Catalog No. 50-291-3698 Supplier Cayman Chemical Supplier No. 3551750 MG
An EGFR inhibitor (Ki = 2.7 nM); selective for EGFR over Src and Abl at 0.1 µM; induces caspase-3 and -7 activity in NCI H358 cells at 1 µM; reduces EGFR autophosphorylation in an HN-5 mouse xenograft model at 25 mg/kg