Learn More
Aviva Systems Biology Corp ERMAP Antibody - middle region (ARP42521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERMAP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ERMAP. Alternate names: MGC118810, MGC118811, MGC118812, MGC118813, PRO2801, RD, SCApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58254