Learn More
Sino Biological Recombinant Human Her2/ERBB2 Protein (GST Tag), Active
Catalog No. 502575066
Encompass_Preferred
Recombinant human HER2 (676-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 50-257-5066 Supplier Sino Biological Supplier No. E2711G500
Recombinant human HER2 (676-end) was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.