Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ERN2, GST Tag Recombinant Protein

Catalog No. A30972
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30972 10 μg
A32890 100 μg
A33375 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A30972 Supplier Invitrogen™ Supplier No. A30972
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This ERN2 recombinant human protein (499-end) was expressed in insect cells. ERN2 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic. The gene accession number for ERN2 is NP_150296.4.

ERN2 serine/threonine kinase or endoplasmic reticulum to nucleus signaling 2 protein is a human homologue of the yeast Ire1 gene product which plays a role in multiple facets of the ER stress response in mammalian cells. This pro-apoptotic appears to play no role in the unfolded-protein response, unlike closely related proteins. Overexpression of ERN2 activates both BiP and CHOP expression, and also leads to the development of programmed cell death. It has been suggested that Ern2 plays a role in multiple facets of the ER stress response in mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q76MJ5
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM EDTA, 150mM NaCl, 0.1mM PMSF, 10mM glutathione and no preservative; pH 7.5
Gene ID (Entrez) 10595
Molecular Weight (g/mol) 79.6 kDa
Name Human ERN2, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias endoplasmic reticulum (ER) to nucleus signalling 2; endoplasmic reticulum to nucleus signaling 2; endoplasmic reticulum to nucleus signalling 2; endoplasmic reticulum-to-nucleus signaling 2; Endoribonuclease; ER to nucleus signalling 2; Ern1; ern2; hIRE2p; inositol-requiring 1 (Yeast homologue); inositol-requiring 1 beta; inositol-requiring protein 2; Ire1; IRE1 beta; IRE1, S. cerevisiae, homolog of; Ire1b; ire1-beta; IRE2; IRE2p; mIre1; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE2
Common Name ERN2
Gene Symbol ERN2
Kinase Family IRE Family
Protein Length 499-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 499-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Active, Catalytic
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.