Learn More
Cayman Chemical Erinacine A, Purity: ≥98%, CAS: 156101-08-5, 5 mg
SDP
Catalog No. 502916269
Encompass_Preferred
A diterpenoid with anticancer and neuroprotective activities; decreases the viability of, and induces cell cycle arrest at the G1 phase and ROS production in, DLD-1 cells at 10 µM; increases the production of NGF in mouse astroglial cells at 1 mM; increases the time spent on the rod in a rotarod test in a mouse model of MPTP-induced Parkinson’s disease at 1 mg/kg, i.p.; oral administration decreases infarct volume in a mouse model of ischemic brain injury induced by transient right common carotid artery occlusion at 4 mg/kg
Catalog No. 50-291-6269 Supplier Cayman Chemical Supplier No. 383525 MG
A diterpenoid with anticancer and neuroprotective activities; decreases the viability of, and induces cell cycle arrest at the G1 phase and ROS production in, DLD-1 cells at 10 µM; increases the production of NGF in mouse astroglial cells at 1 mM; increases the time spent on the rod in a rotarod test in a mouse model of MPTP-induced Parkinson’s disease at 1 mg/kg, i.p.; oral administration decreases infarct volume in a mouse model of ischemic brain injury induced by transient right common carotid artery occlusion at 4 mg/kg