Learn More
Aviva Systems Biology Corp ERP29 Antibody - N-terminal region (ARP44820_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44820_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ERP29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ERP29. Alternate names: C12orf8, ERp28, ERp31, PDI-DB, PDIA9ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53955