Learn More
Enzo Life Sciences ERp57 monoclonal antibody (MaP.Erp57) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA725F
Encompass_Preferred
Clone: MaP.Erp57. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human ERp57. UniProt ID: P30101. Source: Purified from ascites. Species reactivity: Human Porcine, Monkey, Hamster, Dog. Applications: IHC, WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7455