Learn More
Enzo Life Sciences ERp57 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA585F
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat liver ERp57. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Bovine. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7443