Learn More
Sino Biological ERP72 / PDIA4 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 13333RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ERP72 / PDIA4 (rh PDIA4; Catalog#13333-H08H; P13667; Met1-Thr641). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8225