Learn More
Medchemexpress LLC HY-113960 10mg Medchemexpress, ERRα antagonist-1 CAS:1072145-33-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11396010MG
Encompass_Preferred
Medchemexpress, HY-113960 10mg ERRα antagonist-1 CAS:1072145-33-5 ERRα antagonist-1 (Compound A) is a selective and high affinity estrogen-related receptor α (ERRα) antagonist. ERRα antagonist-1 inhibits interaction of ERRα with Proliferator-activated Receptor γ Coactivator-1α (PGC-1α) and PGC-1β, the IC50 values are 170 nM and 180 nM, respectively. ERRα antagonist-1 does not inhibit the interaction of either ERRβ or ERRγ with PGC-1α and PGC-1β coactivator, and also does not inhibit interaction of ERα or ERβ with PGC-1α or SRC-1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9270