Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ERRα ligand-linker conjugates 1 | 00-00-0 | 98.8% | C24H20F9N3O5 | 10MG
SDP

Catalog No. 5000260839
Encompass_Preferred

ERRα ligand-linker conjugate combines an ERRα-targeting ligand with a PROTAC-compatible linker to serve as a building block for synthesis of ERRα-directed degraders and related conjugates. Supplied as a solid with documented purity, molecular properties, and recommended storage for research applications.

  • Designed for use in PROTAC synthesis and conjugation.
  • High purity: 98.8%.
  • Molecular weight: 601.42 g·mol⁻¹.
  • Molecular formula: C24H20F9N3O5.
  • Soluble in DMSO up to 200 mg/mL; ultrasonic may be needed.
  • Appearance: light yellow to yellow solid.
  • Recommended storage: store under nitrogen at -20°C; in solution keep at -80°C for long-term.

Catalog No. 50-002-60839 Supplier Medchemexpress LLC Supplier No. HY13049910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ERRα ligand-linker conjugate combines an ERRα-targeting ligand with a PROTAC-compatible linker to serve as a building block for synthesis of ERRα-directed degraders and related conjugates. Supplied as a solid with documented purity, molecular properties, and recommended storage for research applications.

  • Designed for use in PROTAC synthesis and conjugation.
  • High purity: 98.8%.
  • Molecular weight: 601.42 g·mol⁻¹.
  • Molecular formula: C24H20F9N3O5.
  • Soluble in DMSO up to 200 mg/mL; ultrasonic may be needed.
  • Appearance: light yellow to yellow solid.
  • Recommended storage: store under nitrogen at -20°C; in solution keep at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.