Learn More
Medchemexpress LLC ERRα Ligand-Linker Conjugates 1 | 98.8% | 100 MG
SDP
Catalog No. 5000338727
Encompass_Preferred
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker, which recruits E3 ligases MDM2. This product can be used in the synthesis of a series of PROTACs, such as PROTAC ERRalpha Degrader-1, which is an ERRα degrader.
  • Incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker.
  • Recruits E3 ligases MDM2.
  • Utilized in the synthesis of a series of PROTACs.
  • Appears as a light yellow to yellow solid.
  • Recommended storage at -20°C under nitrogen, away from moisture.
  • High solubility in DMSO (200 mg/mL) with sonication.
Catalog No. 50-003-38727 Supplier Medchemexpress LLC Supplier No. HY130499100MG
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker, which recruits E3 ligases MDM2. This product can be used in the synthesis of a series of PROTACs, such as PROTAC ERRalpha Degrader-1, which is an ERRα degrader.
  • Incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker.
  • Recruits E3 ligases MDM2.
  • Utilized in the synthesis of a series of PROTACs.
  • Appears as a light yellow to yellow solid.
  • Recommended storage at -20°C under nitrogen, away from moisture.
  • High solubility in DMSO (200 mg/mL) with sonication.