Learn More
Medchemexpress LLC ERRα Ligand-Linker Conjugates 1 | 98.8% | 25 MG
SDP
Catalog No. 5000342447
Encompass_Preferred
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker. This compound is designed to recruit E3 ligases MDM2 and can be used in the synthesis of a variety of PROTACs.
  • Incorporates a ligand for ERRα
  • Includes a PROTAC linker
  • Designed to recruit E3 ligases MDM2
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-42447 Supplier Medchemexpress LLC Supplier No. 25MGHY130499
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα) and a PROTAC linker. This compound is designed to recruit E3 ligases MDM2 and can be used in the synthesis of a variety of PROTACs.
  • Incorporates a ligand for ERRα
  • Includes a PROTAC linker
  • Designed to recruit E3 ligases MDM2
  • Used in the synthesis of PROTACs
  • For research use only