Learn More
Medchemexpress LLC ERRα Ligand-Linker Conjugates 1 | 98.8% | 50 MG
SDP
Catalog No. 5000338728
Encompass_Preferred
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα), and a PROTAC linker, which recruit E3 ligases MDM2. It can be used in the synthesis of a series of PROTACs, such as PROTAC ERRalpha Degrader-1. This product is for research use only.
  • Incorporates a ligand for estrogen-related receptor alpha (ERRα)
  • Includes a PROTAC linker
  • Recruits E3 ligases MDM2
  • Can be used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-38728 Supplier Medchemexpress LLC Supplier No. HY13049950MG
ERRα Ligand-Linker Conjugates 1 incorporates a ligand for estrogen-related receptor alpha (ERRα), and a PROTAC linker, which recruit E3 ligases MDM2. It can be used in the synthesis of a series of PROTACs, such as PROTAC ERRalpha Degrader-1. This product is for research use only.
  • Incorporates a ligand for estrogen-related receptor alpha (ERRα)
  • Includes a PROTAC linker
  • Recruits E3 ligases MDM2
  • Can be used in the synthesis of PROTACs
  • For research use only