Learn More
Medchemexpress LLC ERRα ligand-linker conjugates 1 | 98.8% | 601.42 | C24H20F9N3O5 | 5 MG
SDP
Catalog No. 5000262882
Encompass_Preferred
ERRα Ligand-linker conjugates 1 is a small-molecule ligand-linker intended as a building block for PROTAC design against estrogen-related receptor alpha (ERRα). It combines an ERRα-binding ligand with a PROTAC-compatible linker to facilitate recruitment of E3 ligases and construction of ERRα degraders. The material is supplied as a solid for medicinal chemistry and degradation studies.
  • Designed for PROTAC synthesis
  • Incorporates an ERRα-targeting ligand
  • Compatible with common E3 ligase recruiters
  • High purity suitable for research (98.8%)
  • Supplied as a 5 mg solid
  • Characterized: molecular weight 601.42, formula C24H20F9N3O5
Catalog No. 50-002-62882 Supplier Medchemexpress LLC Supplier No. HY1304995MG
ERRα Ligand-linker conjugates 1 is a small-molecule ligand-linker intended as a building block for PROTAC design against estrogen-related receptor alpha (ERRα). It combines an ERRα-binding ligand with a PROTAC-compatible linker to facilitate recruitment of E3 ligases and construction of ERRα degraders. The material is supplied as a solid for medicinal chemistry and degradation studies.
  • Designed for PROTAC synthesis
  • Incorporates an ERRα-targeting ligand
  • Compatible with common E3 ligase recruiters
  • High purity suitable for research (98.8%)
  • Supplied as a 5 mg solid
  • Characterized: molecular weight 601.42, formula C24H20F9N3O5