Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Erso | 2407860-35-7 | 100.0% | C22H13F6NO3 | 10MG
SDP

Catalog No. 5000261855
Encompass_Preferred

ErSO is a small-molecule selective anticipatory unfolded protein response (a-UPR) activator that engages estrogen receptor alpha (ERα) to elicit potent and sustained cytotoxic a-UPR signaling in ERα-positive cancer models. It is supplied for research use and characterized by a defined molecular formula and high analytical purity.

  • Activates the anticipatory unfolded protein response (a-UPR) via ERα.
  • Induces potent and sustained cytotoxic signaling in ERα-positive models.
  • Suitable for cancer research applications.
  • High analytical purity reported at 99.95%.
  • Molecular formula C22H13F6NO3; molecular weight 453.33 g/mol.
  • Available in small research pack sizes (for example, 10 mg).

Catalog No. 50-002-61855 Supplier Medchemexpress LLC Supplier No. HY13224710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ErSO is a small-molecule selective anticipatory unfolded protein response (a-UPR) activator that engages estrogen receptor alpha (ERα) to elicit potent and sustained cytotoxic a-UPR signaling in ERα-positive cancer models. It is supplied for research use and characterized by a defined molecular formula and high analytical purity.

  • Activates the anticipatory unfolded protein response (a-UPR) via ERα.
  • Induces potent and sustained cytotoxic signaling in ERα-positive models.
  • Suitable for cancer research applications.
  • High analytical purity reported at 99.95%.
  • Molecular formula C22H13F6NO3; molecular weight 453.33 g/mol.
  • Available in small research pack sizes (for example, 10 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.