Learn More
Medchemexpress LLC ErSO-DFP | 2768139-76-8 | 99.6% | C20H17F5N2O2 | 50 MG
SDP
Catalog No. 5000355083
Encompass_Preferred
ErSO-DFP is an anticipatory unfolded protein response (a-UPR) activator. It demonstrates enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells, offering a wider selectivity window. This compound exhibits antitumor activity against MCF-7 in mice and can effectively cross the blood-brain barrier.
  • Anticipatory unfolded protein response (a-UPR) activator
  • Enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells
  • Wider selectivity window
  • Displays antitumor activity against MCF-7 in mice
  • Can cross the blood-brain barrier
Catalog No. 50-003-55083 Supplier Medchemexpress LLC Supplier No. HY14407050MG
ErSO-DFP is an anticipatory unfolded protein response (a-UPR) activator. It demonstrates enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells, offering a wider selectivity window. This compound exhibits antitumor activity against MCF-7 in mice and can effectively cross the blood-brain barrier.
  • Anticipatory unfolded protein response (a-UPR) activator
  • Enhanced selectivity for estrogen receptor alpha-positive (ERα+) cancer cells
  • Wider selectivity window
  • Displays antitumor activity against MCF-7 in mice
  • Can cross the blood-brain barrier