Learn More
Medchemexpress LLC Ertapenem disodium | 153832-38-3 | 97.5% | C22H23N3Na2O7S | 5 MG
SDP
Catalog No. 5000432909
Encompass_Preferred
Ertapenem disodium is a broad-spectrum and long-acting β-lactam antibiotic. This compound demonstrates broad-spectrum anti-anaerobic activity, with effectiveness against 99.1% of anaerobes, exhibiting a mode MIC of 0.12 μg/mL and an MIC90 of 1 μg/mL. It is utilized in research for severe infections caused by bacteria in various bodily locations, including the skin, lungs, stomach, pelvis, and urinary tract. The compound is active against both gram-positive and gram-negative organisms.
  • Shipping at room temperature within the continental US.
  • General storage: -20°C, in a sealed container, protected from moisture and light.
  • In solvent (stock solution): -80°C for up to 6 months or -20°C for up to 1 month.
  • For both in solvent conditions, ensure sealed storage, away from moisture and light.
  • If water is used as stock solution, dilute to working solution, filter and sterilize with a 0.22 μm filter before use.
Catalog No. 50-004-32909 Supplier Medchemexpress LLC Supplier No. HYA0294A5MG
Ertapenem disodium is a broad-spectrum and long-acting β-lactam antibiotic. This compound demonstrates broad-spectrum anti-anaerobic activity, with effectiveness against 99.1% of anaerobes, exhibiting a mode MIC of 0.12 μg/mL and an MIC90 of 1 μg/mL. It is utilized in research for severe infections caused by bacteria in various bodily locations, including the skin, lungs, stomach, pelvis, and urinary tract. The compound is active against both gram-positive and gram-negative organisms.
  • Shipping at room temperature within the continental US.
  • General storage: -20°C, in a sealed container, protected from moisture and light.
  • In solvent (stock solution): -80°C for up to 6 months or -20°C for up to 1 month.
  • For both in solvent conditions, ensure sealed storage, away from moisture and light.
  • If water is used as stock solution, dilute to working solution, filter and sterilize with a 0.22 μm filter before use.