Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ertapenem disodium | 153832-38-3 | 98.2% | 519.48 | C22H23N3Na2O7S | 10 MG
SDP

Catalog No. 5000298393
Encompass_Preferred

Ertapenem disodium is the disodium salt of ertapenem, a long-acting, broad-spectrum carbapenem β-lactam antibiotic supplied as a high-purity, research-grade powder for in vitro and in vivo studies of bacterial infection. It is used in microbiology and pharmacology research to evaluate antibacterial activity and pharmacokinetics.

  • Purity 98.21%.
  • CAS number 153832-38-3.
  • Molecular formula C22H23N3Na2O7S and molecular weight 519.48.
  • Supplied as a powder; common unit size 10 MG.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-98393 Supplier Medchemexpress LLC Supplier No. HYA0294A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ertapenem disodium is the disodium salt of ertapenem, a long-acting, broad-spectrum carbapenem β-lactam antibiotic supplied as a high-purity, research-grade powder for in vitro and in vivo studies of bacterial infection. It is used in microbiology and pharmacology research to evaluate antibacterial activity and pharmacokinetics.

  • Purity 98.21%.
  • CAS number 153832-38-3.
  • Molecular formula C22H23N3Na2O7S and molecular weight 519.48.
  • Supplied as a powder; common unit size 10 MG.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.