Learn More
Medchemexpress LLC Ertapenem sodium | 153773-82-1 | >98.2% | 497.50 | 500 MG
SDP
Catalog No. 5000349743
Encompass_Preferred
Ertapenem sodium (L-749345) is a broad-spectrum and long-acting β-lactam antibiotic. It exhibits broad-spectrum anti-anaerobic activity against various anaerobes with a mode MIC of 0.12 μg/mL. This product can be used for research on severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • Broad-spectrum, long-acting β-lactam antibiotic
  • Effective against various anaerobes
  • Useful for research on severe bacterial infections
Catalog No. 50-003-49743 Supplier Medchemexpress LLC Supplier No. HY13625500MG
Ertapenem sodium (L-749345) is a broad-spectrum and long-acting β-lactam antibiotic. It exhibits broad-spectrum anti-anaerobic activity against various anaerobes with a mode MIC of 0.12 μg/mL. This product can be used for research on severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • Broad-spectrum, long-acting β-lactam antibiotic
  • Effective against various anaerobes
  • Useful for research on severe bacterial infections