Learn More
Medchemexpress LLC Ertapenem sodium | 153773-82-1 | MFCD09475566 | 98.9% | 497.50 | C22H24N3NaO7S | 5 MG
SDP
Catalog No. 5000222203
Encompass_Preferred
Ertapenem sodium is a research-grade carbapenem antibiotic provided as a solid analytical standard for laboratory use. It is intended for antimicrobial research, susceptibility testing, and analytical method development, and is supplied in small measured quantities with documented purity and physicochemical data.
Catalog No. 50-002-22203 Supplier Medchemexpress LLC Supplier No. HY136255MG
Ertapenem sodium is a research-grade carbapenem antibiotic provided as a solid analytical standard for laboratory use. It is intended for antimicrobial research, susceptibility testing, and analytical method development, and is supplied in small measured quantities with documented purity and physicochemical data.