Learn More
Selleck Chemical LLC Ertapenem sodium
SDP
Supplier:  Selleck Chemical LLC S565725MG
Encompass_Preferred
Ertapenem Sodium (MK826) is the sodium salt of ertapenem a long-acting broad-spectrum antibiotic of -lactam subclass
Catalog No. 50-000-03699