Learn More
TARGETMOL CHEMICALS INC ERTAPENEM SODIUM 50MG
SDP
Catalog No. 502576727
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ertapenem sodium (MK-826) is a new long-acting 1-(beta)-methyl carbapenem antibiotic with a broad antibacterial spectrum including common aerobic and anaerobic bacteria and organisms with extended-spectrum (beta)-lactamases. purity: 96%
Catalog No. 50-257-6727 Supplier TARGETMOL CHEMICALS INC Supplier No. T499150MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ertapenem sodium (MK-826) is a new long-acting 1-(beta)-methyl carbapenem antibiotic with a broad antibacterial spectrum including common aerobic and anaerobic bacteria and organisms with extended-spectrum (beta)-lactamases. purity: 96%