Learn More
Research Products International Corp Ertapenem Sodium Salt, 5 Grams
SDP
Catalog No. 50997777
Encompass_Preferred
Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.
It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria, including multidrug-resistant strains.
In the field of microbiology and infectious disease research, Ertapenem Sodium Salt is commonly used as a tool for studying bacterial susceptibility and resistance. It can be employed in antimicrobial susceptibility testing to determine the efficacy of different antibiotics against bacterial isolates.
Catalog No. 50-997-777 Supplier Research Products International Corp Supplier No. E321005.0
Ertapenem Sodium Salt is a potent antibiotic and belongs to the carbapenem class of antibiotics.
It is effective against a broad spectrum of Gram-positive and Gram-negative bacteria, including multidrug-resistant strains.
In the field of microbiology and infectious disease research, Ertapenem Sodium Salt is commonly used as a tool for studying bacterial susceptibility and resistance. It can be employed in antimicrobial susceptibility testing to determine the efficacy of different antibiotics against bacterial isolates.