Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ertapenem sodium (Standard) | 153773-82-1 | 97.39% | 497.50 | 100 MG
SDP

Catalog No. 5000349574
Encompass_Preferred

Ertapenem sodium (Standard) is an analytical standard and a broad-spectrum, long-acting β-lactam antibiotic. It is primarily used for research and analytical applications, including qualitative, quantitative, and methodological experiments like HPLC, GC, and MS. This compound is also studied for its efficacy against severe bacterial infections affecting the skin, lungs, stomach, pelvis, and urinary tract.

  • Broad-spectrum and long-acting β-lactam antibiotic
  • Exhibits anti-anaerobic activity with a mode MIC of 0.12 μg/mL
  • Ideal for HPLC, GC, and MS research
  • 97.39% purity
  • Molecular weight: 497.50
  • White to off-white solid appearance
  • Recommended storage at -20°C, sealed, away from light and moisture

Catalog No. 50-003-49574 Supplier Medchemexpress LLC Supplier No. HY13625R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ertapenem sodium (Standard) is an analytical standard and a broad-spectrum, long-acting β-lactam antibiotic. It is primarily used for research and analytical applications, including qualitative, quantitative, and methodological experiments like HPLC, GC, and MS. This compound is also studied for its efficacy against severe bacterial infections affecting the skin, lungs, stomach, pelvis, and urinary tract.

  • Broad-spectrum and long-acting β-lactam antibiotic
  • Exhibits anti-anaerobic activity with a mode MIC of 0.12 μg/mL
  • Ideal for HPLC, GC, and MS research
  • 97.39% purity
  • Molecular weight: 497.50
  • White to off-white solid appearance
  • Recommended storage at -20°C, sealed, away from light and moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.