Learn More
Medchemexpress LLC Ertapenem sodium (Standard) | 153773-82-1 | 97.4% | 497.50 | 50 MG
SDP
Catalog No. 5000352135
Encompass_Preferred
Ertapenem sodium (Standard) is the analytical standard for Ertapenem sodium. This broad-spectrum β-lactam antibiotic exhibits strong anti-anaerobic activity and is intended for research and analytical applications. It is used for research on severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • Intended for research and analytical applications
  • Broad-spectrum and long-acting β-lactam antibiotic
  • Exhibits broad-spectrum anti-anaerobic activity
  • Used for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-52135 Supplier Medchemexpress LLC Supplier No. HY13625R50MG
Ertapenem sodium (Standard) is the analytical standard for Ertapenem sodium. This broad-spectrum β-lactam antibiotic exhibits strong anti-anaerobic activity and is intended for research and analytical applications. It is used for research on severe infections caused by bacteria in the skin, lungs, stomach, pelvis, and urinary tract.
  • Intended for research and analytical applications
  • Broad-spectrum and long-acting β-lactam antibiotic
  • Exhibits broad-spectrum anti-anaerobic activity
  • Used for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS