Learn More
Medchemexpress LLC HY-18295 1mg Medchemexpress, Erteberel CAS:533884-09-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY182951MG
Encompass_Preferred
Medchemexpress, HY-18295 1mg Erteberel CAS:533884-09-2 Erteberel (LY500307) is a potent and selective estrogen receptor beta (ERβ) agonist with Ki and EC50 of 1.54 nM and 3.61 nM, respectively. Anti-tumor activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0003