Learn More
TARGETMOL CHEMICALS INC ERTEBEREL 5MG
SDP
Catalog No. 502579442
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Erteberel (LY500307) is a potent selective estrogen receptor (beta) agonist with EC50 of 0.66 nM 32-fold selectivity against estrogen receptor (alpha). Phase 2. purity: 99%
Catalog No. 50-257-9442 Supplier TARGETMOL CHEMICALS INC Supplier No. T63065MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Erteberel (LY500307) is a potent selective estrogen receptor (beta) agonist with EC50 of 0.66 nM 32-fold selectivity against estrogen receptor (alpha). Phase 2. purity: 99%