Learn More
Selleck Chemical LLC Ertugliflozin S5413-25mg
SDP
Catalog No. 503109515
Encompass_Preferred
Ertugliflozin (MK-8835 PF-04971729) is a potent and selective sodium-dependent glucose cotransporter 2 inhibitor with IC50 values of 0 877 nM for h-SGLT2 and 1000-fold higher for h-SGLT1
Catalog No. 50-310-9515 Supplier Selleck Chemical LLC Supplier No. S541325MG
Ertugliflozin (MK-8835 PF-04971729) is a potent and selective sodium-dependent glucose cotransporter 2 inhibitor with IC50 values of 0 877 nM for h-SGLT2 and 1000-fold higher for h-SGLT1