Learn More
Cayman Chemical Ertugliflozin, Purity: ≥98%, CAS: 1210344-57-2, 25 mg
SDP
Catalog No. 502964025
Encompass_Preferred
An SGLT2 inhibitor (IC50 = 0.927 nM for the human enzyme); selective for SGLT2 over SGLT1 (IC50 = 2,050 nM for the human enzyme); increases urinary glucose excretion in rats in a dose-dependent manner
Catalog No. 50-296-4025 Supplier Cayman Chemical Supplier No. 2150725 MG
An SGLT2 inhibitor (IC50 = 0.927 nM for the human enzyme); selective for SGLT2 over SGLT1 (IC50 = 2,050 nM for the human enzyme); increases urinary glucose excretion in rats in a dose-dependent manner